accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1120-24-7

IUPAC Name: decyldimethylamine
Synonyms: decyldimethylamine
Molecular Weight (g/mol): 185.36
Molecular Formula: C12H27N
InChi Key: YWWNNLPSZSEZNZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCN(C)C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)