accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112147-27-0

IUPAC Name: disodium 3-[(3,5-dibromo-4-oxidophenyl)imino]-6-oxocyclohexa-1,4-diene-1-carboxylate
Synonyms: disodium 3-[(3,5-dibromo-4-oxidophenyl)imino]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol): 444.97
Molecular Formula: C13H5Br2NNa2O4
InChi Key: BHCDHBWWPPYBTK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC(C=CC1=O)=NC1=CC(Br)=C([O-])C(Br)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results