Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromoindosalicylic Acid Disodium Salt, TCI America™
SDP

Catalog No. D01911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D01911G Supplier TCI America Supplier No. D01911G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112147-27-0
Molecular Formula C13H5Br2NNa2O4
Molecular Weight (g/mol) 444.974
MDL Number MFCD00059574
InChI Key BHCDHBWWPPYBTK-UHFFFAOYSA-L
Synonym 2,6-Dibromophenolindosalicylic Acid Disodium Salt
PubChem CID 44630222
IUPAC Name disodium;3-(3,5-dibromo-4-oxidophenyl)imino-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES C1=CC(=O)C(=CC1=NC2=CC(=C(C(=C2)Br)[O-])Br)C(=O)[O-].[Na+].[Na+]

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 444.97
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromoindosalicylic Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.