accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNHNNH

CAS RN 112193-83-6

IUPAC Name: 1-benzyl-1,4,7,10-tetraazacyclododecane
Synonyms: 1-benzyl-1,4,7,10-tetraazacyclododecane
Molecular Weight (g/mol): 262.4
Molecular Formula: C15H26N4
InChi Key: FURLCQRFFWBENR-UHFFFAOYSA-N
SMILES: C(N1CCNCCNCCNCC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results