accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSN

CAS RN 1122-83-4

IUPAC Name: N-(oxo-λ⁴-sulfanylidene)aniline
Synonyms: N-sulfinylaniline
Molecular Weight (g/mol): 139.17
Molecular Formula: C6H5NOS
InChi Key: FIOJWGRGPONADF-UHFFFAOYSA-N
SMILES: O=S=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)