Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Thionylaniline

Catalog No. T3170425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3170425G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3170425G Supplier Sigma Aldrich Supplier No. T3170425G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (1-bromoethyl)benzene has been employed:
  • In controlled radical polymerization of styrene
  • In asymmetric esterification of benzoic acid in the presence of a chiral cyclic guanidine
  • As initiator in the synthesis of bromine terminated polyp-methoxystyrene and polystyrene via atom transfer radical polymerization

Specifications

CAS 1122-83-4
Density 1.236 g/mL (at 25°C (literature))
Boiling Point 200°C (lit.), 83°C (15 mmHg)
Molecular Formula C6H5NOS
Refractive Index n20/D 1.627 (literature)
Linear Formula C6H5NSO
MDL Number MFCD00002084
Quantity 100 g
Synonym N-Sulfinyl-aniline
Molecular Weight (g/mol) 139.18
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.