accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHNH2

CAS RN 112259-66-2

IUPAC Name: 2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
Synonyms: 2-amino-4-(cyclohexyloxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: IJAPBBOAGRLKMH-UHFFFAOYNA-N
SMILES: NC(CC(=O)OC1CCCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)