accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1124-16-9

IUPAC Name: 3-methyl-1-(propan-2-yl)-1H-pyrazol-5-amine
Synonyms: 2-isopropyl-5-methylpyrazol-3-amine
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: OCSWHADWFJTNAZ-UHFFFAOYSA-N
SMILES: CC(C)N1N=C(C)C=C1N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results