Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-isopropyl-5-methyl-2H-pyrazole, 97%, Thermo Scientific™

Catalog No. AAH6674206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6674206 5 g
AAH6674203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6674206 Supplier Thermo Scientific Chemicals Supplier No. H6674206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-16-9
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
MDL Number MFCD00514520
InChI Key OCSWHADWFJTNAZ-UHFFFAOYSA-N
Synonym 1-isopropyl-3-methyl-1h-pyrazol-5-amine, 5-amino-1-isopropyl-3-methylpyrazole, 2-isopropyl-5-methyl-2h-pyrazol-3-ylamine, 2-isopropyl-5-methylpyrazol-3-amine, 1h-pyrazol-5-amine, 3-methyl-1-1-methylethyl, pyrazole, 5-amino-1-isopropyl-3-methyl, 3-methyl-1-propan-2-yl-1h-pyrazol-5-amine, 3-methyl-1-methylethyl pyrazole-5-ylamine, 5-25-09-00465 beilstein handbook reference, 1-isopropyl-3-methyl-5-aminopyrazol
PubChem CID 121018
IUPAC Name 5-methyl-2-propan-2-ylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C(C)C

Specifications

Quantity 5 g
Beilstein 607365
Formula Weight 139.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Amino-2-isopropyl-5-methyl-2H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ5778000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.