accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1124-62-5

IUPAC Name: (3-chloropropyl)cyclohexane
Synonyms: (3-chloropropyl)cyclohexane
Molecular Weight (g/mol): 160.69
Molecular Formula: C9H17Cl
InChi Key: QEISABAAOUXQNG-UHFFFAOYSA-N
SMILES: ClCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results