Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyclohexylpropyl Chloride 99+%, Thermo Scientific™

Catalog No. AC255970250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC255970250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255970250 Supplier Thermo Scientific Chemicals Supplier No. 255970250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-62-5
Molecular Formula C9H17Cl
Molecular Weight (g/mol) 160.685
InChI Key QEISABAAOUXQNG-UHFFFAOYSA-N
Synonym 3-chloropropyl cyclohexane, 1-chloro-3-cyclohexylpropane, cyclohexane, 3-chloropropyl, chloropropylcyclohexane, 3-cyclohexylpropyl chloride, 3-cyclohexyl-1-chloropropane
PubChem CID 70762
IUPAC Name 3-chloropropylcyclohexane
SMILES C1CCC(CC1)CCCCl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4650 to 1.4670 (20°C, 589nm)
Linear Formula C6H11(CH2)3Cl
Quantity 25 mL
Formula Weight 160.59
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 3-Cyclohexylpropyl chloride, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.