accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112465-94-8

IUPAC Name: hydrogen 3,4-dichloro-N-methyl-N-[(1R,2S)-2-(pyrrolidin-1-yl)cyclohexyl]benzamide chloride
Synonyms: hydrogen 3,4-dichloro-N-methyl-N-[(1R,2S)-2-(pyrrolidin-1-yl)cyclohexyl]benzamide chloride
Molecular Weight (g/mol): 391.76
Molecular Formula: C18H25Cl3N2O
InChi Key: WFUASZXAHZXJMX-PPPUBMIESA-N
SMILES: [H+].[Cl-].CN([C@@H]1CCCC[C@@H]1N1CCCC1)C(=O)C1=CC=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results