accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3O

CAS RN 1125-21-9

IUPAC Name: 2,6,6-trimethylcyclohex-2-ene-1,4-dione
Synonyms: 2,6,6-trimethylcyclohex-2-ene-1,4-dione
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: AYJXHIDNNLJQDT-UHFFFAOYSA-N
SMILES: CC1=CC(=O)CC(C)(C)C1=O
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)