Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6,6-Trimethyl-2-cyclohexene-1,4-dione, 98%, Thermo Scientific™

Catalog No. AC334531000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC334531000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC334531000 Supplier Thermo Scientific Chemicals Supplier No. 334531000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1125-21-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00043119
InChI Key AYJXHIDNNLJQDT-UHFFFAOYSA-N
Synonym 4-oxoisophorone, 2,6,6-trimethyl-2-cyclohexene-1,4-dione, ketoisophorone, oxopholone, keto-isophorone, 4-ketoisophorone, oxophorone, 6-oxoisophorone, 2-cyclohexene-1,4-dione, 2,6,6-trimethyl, 4-oxo-alpha-isophorone
PubChem CID 62374
IUPAC Name 2,6,6-trimethylcyclohex-2-ene-1,4-dione
SMILES CC1=CC(=O)CC(C)(C)C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Orange
Density 1.03g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
Quantity 100 g
Beilstein 7, IV, 2032
Formula Weight 152.19
Percent Purity ≥97.5%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2,6,6-Trimethyl-2-cyclohexene-1,4-dione, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.