accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3OOH

CAS RN 1125-29-7

IUPAC Name: trimethyl-1H-pyrazole-4-carboxylic acid
Synonyms: trimethylpyrazole-4-carboxylic acid
Molecular Weight (g/mol): 154.17
Molecular Formula: C7H10N2O2
InChi Key: NOIOGQJFLIPRBI-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(C(O)=O)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results