Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC14701EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC14701EA 10 g Amber Glass Bottle
CC14701CB 250 mg Amber Glass Bottle
CC14701DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC14701EA Supplier Thermo Scientific Chemicals Supplier No. CC14701EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1125-29-7
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD00159648
InChI Key NOIOGQJFLIPRBI-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole-4-carboxylic acid, trimethyl-1h-pyrazole-4-carboxylic acid, trimethylpyrazole-4-carboxylic acid, 1,3,5-trimethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1,3,5-trimethyl, akos pao-0337, rarechem al be 0403, art-chem-bb b006363, cambridge id 5264152, 1,3,5-trimethyl-1h-pyrazol-4-carboxylic acid
PubChem CID 736514
IUPAC Name 1,3,5-trimethylpyrazole-4-carboxylic acid
SMILES CN1N=C(C)C(C(O)=O)=C1C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazole-4-carboxylic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.