accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COCH3

CAS RN 1126-93-8

IUPAC Name: 2-ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 172.03
Molecular Formula: C8H17BO3
InChi Key: AWCHCIVACATJJS-UHFFFAOYSA-N
SMILES: CCOB1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results