Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
SDP

Catalog No. E09825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09825G 5 g
E098225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09825G Supplier TCI America Supplier No. E09825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1126-93-8
Molecular Formula C8H17BO3
Molecular Weight (g/mol) 172.03
MDL Number MFCD22054335
InChI Key AWCHCIVACATJJS-UHFFFAOYSA-N
Synonym Ethoxyboronic Acid Pinacol Ester
PubChem CID 12727575
IUPAC Name 2-ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCOB1OC(C)(C)C(C)(C)O1

Specifications

Color Colorless
Boiling Point 32°C
Quantity 5 g
UN Number 1993
Formula Weight 172.03
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.