accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112626-50-3

NHHCl

CAS RN 112626-50-3

IUPAC Name: octahydrocyclopenta[c]pyrrole hydrochloride
Synonyms: octahydrocyclopenta[c]pyrrole hydrochloride
Molecular Weight (g/mol): 147.65
Molecular Formula: C7H14ClN
InChi Key: HVZRRRPCVOJOLJ-UHFFFAOYNA-N
SMILES: Cl.C1CC2CNCC2C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)