Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Azabicyclo[3.3.0]octane hydrochloride, 99%, Thermo Scientific™

Catalog No. AC457701000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457701000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC457701000 Supplier Thermo Scientific Chemicals Supplier No. 457701000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112626-50-3
Molecular Formula C7H13N·ClH
Molecular Weight (g/mol) 147.65
InChI Key HVZRRRPCVOJOLJ-UHFFFAOYSA-N
Synonym 3-azabicyclo 3.3.0 octane hydrochloride, octahydrocyclopenta c pyrrole hydrochloride, 3-azabicyclo 3.3.0 octane hcl, 3-azabicyclo 3,3,0 octane hcl, 3-azabicyclo 3.3.0 octanehydrochloride, 1,2,3,3a,4,5,6,6a-octahydrocyclopenta c pyrrole hydrochloride, pubchem9779, cis-octahydro-cyclopenta c pyrrole hydrochloride, c7h14ncl, 3-azabicyclo 3.3.0 octane hydrochloride;
PubChem CID 21572123
IUPAC Name 1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
SMILES C1CC2CNCC2C1.Cl

Specifications

Melting Point 116.0°C to 118.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 to max. 101.5% min. (Argentometry)
Quantity 100 g
Formula Weight 147.65
Percent Purity 99%
Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.