accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112803-88-0

HClOHOONH2NH(S)

CAS RN 112803-88-0

IUPAC Name: benzyl N-[(5S)-5-amino-6-hydroxyhexyl]carbamate hydrochloride
Synonyms: benzyl N-[(5S)-5-amino-6-hydroxyhexyl]carbamate hydrochloride
Molecular Weight (g/mol): 302.8
Molecular Formula: C14H23ClN2O3
InChi Key: FCQMQDFFGAYKMW-ZOWNYOTGSA-N
SMILES: Cl.N[C@H](CO)CCCCNC(=O)OCC1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)