accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNOOH

CAS RN 112811-90-2

IUPAC Name: 2-(benzylsulfanyl)pyridine-3-carboxylic acid
Synonyms: 2-(benzylsulfanyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 245.3
Molecular Formula: C13H11NO2S
InChi Key: OJNZDGDYAXCHPB-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(SCC2=CC=CC=C2)N=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results