Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Benzylthio)nicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. B40885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40885G 5 g
B40881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40885G Supplier TCI America Supplier No. B40885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 112811-90-2 | C13H11NO2S | 245.296 g/mol

Chemical Identifiers

CAS 112811-90-2
Molecular Formula C13H11NO2S
Molecular Weight (g/mol) 245.296
MDL Number MFCD00661380
InChI Key OJNZDGDYAXCHPB-UHFFFAOYSA-N
Synonym 2-(Benzylthio)pyridine-3-carboxylic Acid
PubChem CID 757779
IUPAC Name 2-benzylsulfanylpyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)CSC2=C(C=CC=N2)C(=O)O

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 5 g
Formula Weight 245.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Benzylthio)nicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.