accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNOO

CAS RN 113-48-4

IUPAC Name: 4-(2-ethylhexyl)-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: ethylhexylbicycloheptene dicarboximide
Molecular Weight (g/mol): 275.39
Molecular Formula: C17H25NO2
InChi Key: WLLGXSLBOPFWQV-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C(=O)C2C3CC(C=C3)C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results