accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2NN

CAS RN 1131-18-6

IUPAC Name: 3-methyl-1-phenyl-1H-pyrazol-5-amine
Synonyms: 5-methyl-2-phenylpyrazol-3-amine
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: FMKMKBLHMONXJM-UHFFFAOYSA-N
SMILES: CC1=NN(C(N)=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)