Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-3-methyl-1-phenylpyrazole, 97%

Catalog No. AC449600250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449600250 25 g
AC449600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449600250 Supplier Thermo Scientific Chemicals Supplier No. 449600250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1131-18-6 | C10H11N3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-18-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.21
InChI Key FMKMKBLHMONXJM-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
PubChem CID 70801
IUPAC Name 5-methyl-2-phenylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C2=CC=CC=C2

Specifications

Melting Point 113°C to 115°C
Boiling Point 333°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 173.21
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 5-Amino-3-methyl-1-phenylpyrazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H StatementHarmful if swallowed.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Precautionary Statement GHS P StatementAvoid breathing dust/fume/gas/mist/vapors/spray.IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.IF ON SKIN: Wash with plenty of soap and water.Wear protective gloves/protective clothing/e
EINECSNumber 214-463-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.