CAS RN 113451-59-5
(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%
CAS: 113451-59-5 Molecular Formula: C10H18N2O2 Molecular Weight (g/mol): 198.26 MDL Number: MFCD01569250 InChI Key: UXAWXZDXVOYLII-YUMQZZPRSA-N Synonym: 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane,1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate,1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane,1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate PubChem CID: 11521263 IUPAC Name: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate SMILES: CC(C)(C)OC(=O)N1CC2CC1CN2
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%
CAS: 113451-59-5 Molecular Formula: C10H18N2O2 Molecular Weight (g/mol): 198.266 MDL Number: MFCD01569250 InChI Key: UXAWXZDXVOYLII-YUMQZZPRSA-N Synonym: 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane,1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate,1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane,1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate PubChem CID: 11521263 IUPAC Name: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate SMILES: CC(C)(C)OC(=O)N1CC2CC1CN2
Sigma Aldrich Fmoc-Ala-Wang Resin
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.