Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 98%, Thermo Scientific™

Catalog No. AAH51097MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51097MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH51097MD Supplier Thermo Scientific Chemicals Supplier No. H51097MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 113451-59-5
Molecular Formula C10H18N2O2
Quantity 250mg
MDL Number MFCD01569250
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
Optical Rotation −44° (c=1 in Chloroform)
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
Molecular Weight (g/mol) 198.266
PubChem CID 11521263
Formula Weight 198.26
Percent Purity 98%
Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.