CAS RN 1135256-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ EGF/FGF/PDGF Receptor Tyrosine Kinase Inhibitor
The EGF/FGF/PDGF Receptor Tyrosine Kinase Inhibitor controls the biological activity of EGF/FGF/PDGF Receptor Tyrosine Kinase. Primarily used for Phosphorylation & Dephosphorylation applications.