Learn More
MilliporeSigma™ Calbiochem™ EGF/FGF/PDGF Receptor Tyrosine Kinase Inhibitor
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Color | Pale Yellow |
| Description | 97% by HPLC |
| CAS | 1135256-66-4 |
| Molecular Formula | C18H18Cl2N6O |
| Formula Weight | 405.3g/mol |
| Solubility | DMSO or EtOH |
| Content And Storage | Protect from light |
| Form | Solid |
| Quantity | 2 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.