accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1135616-49-7

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-[(Z)-N''-methyl-N'-[(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-yl)sulfonyl]carbamimidamido]pentanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-[(Z)-N''-methyl-N'-(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-ylsulfonyl)carbamimidamido]pentanoic acid
Molecular Weight (g/mol): 662.8
Molecular Formula: C35H42N4O7S
InChi Key: JAUPJPAADXVUGQ-LJAQVGFWSA-N
SMILES: C\N=C(\NCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)NS(=O)(=O)C1=C(C)C2=C(OC(C)(C)C2)C(C)=C1C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)