accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113571-15-6

IUPAC Name: 4,6-dibromo-2,3-dichloroaniline
Synonyms: 4,6-dibromo-2,3-dichloroaniline
Molecular Weight (g/mol): 319.81
Molecular Formula: C6H3Br2Cl2N
InChi Key: IPWGDMQXLGFBLC-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(Cl)=C(Br)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results