accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOPOOOOHOHOPOOONaNaNaNa

CAS RN 113599-17-0

IUPAC Name: tetrasodium 3,4-dihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl phosphate
Synonyms: tetrasodium 3,4-dihydroxy-5-[(phosphonatooxy)methyl]oxolan-2-yl phosphate
Molecular Weight (g/mol): 398.02
Molecular Formula: C5H8Na4O11P2
InChi Key: NOGFWTMOJJPOTG-UHFFFAOYNA-J
SMILES: [Na+].[Na+].[Na+].[Na+].OC1C(O)C(OP([O-])([O-])=O)OC1COP([O-])([O-])=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results