accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClOH2NFHCl

CAS RN 113674-95-6

IUPAC Name: 4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline hydrochloride
Synonyms: 4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluoroaniline hydrochloride
Molecular Weight (g/mol): 342.11
Molecular Formula: C13H9Cl2F4NO
InChi Key: BBBUNESZGCZWJR-UHFFFAOYSA-N
SMILES: Cl.NC1=C(F)C=C(OC2=C(Cl)C=C(C=C2)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results