accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHH3CH3CCH3

CAS RN 1138-52-9

IUPAC Name: 3,5-di-tert-butylphenol
Synonyms: 3,5-di-T-butylphenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: ZDWSNKPLZUXBPE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=CC(O)=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results