Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Di-Tert-Butylphenol, MP Biomedicals™

Catalog No. MP520509810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP520509810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP520509810 Supplier MP Biomedicals, Inc Supplier No. 0520509810
Add to Cart
Add to Cart

3,5-Di-Tert-Butylphenol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1138-52-9
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00008829
InChI Key ZDWSNKPLZUXBPE-UHFFFAOYSA-N

Specifications

Melting Point 87°C to 89°C (literature)
Color Gray
Quantity 10 g
Physical Form Powder
Grade 0.98
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.