accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114-76-1

IUPAC Name: sodium 2-oxo-3-phenylpropanoate
Synonyms: sodium β-phenylpyruvate
Molecular Weight (g/mol): 186.14
Molecular Formula: C9H7NaO3
InChi Key: MQGYVGKMCRDEAF-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)