Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phenylpyruvate Monohydrate, TCI America™
SDP

Catalog No. P08385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08385G 5 g
P083825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08385G Supplier TCI America Supplier No. P08385G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 114-76-1
Molecular Formula C9H7NaO3
Molecular Weight (g/mol) 186.142
MDL Number MFCD00002590
InChI Key MQGYVGKMCRDEAF-UHFFFAOYSA-M
Synonym sodium phenylpyruvate, phenylpyruvic acid sodium salt, sodium 2-oxo-3-phenylpropanoate, unii-7528cw33ww, sodium ?-phenylpyruvate, phenylpyruvic acid, sodium salt, l-phenyl pyruvic acid sodium salt, beta-phenylpyruvic acid sodium salt, 2-oxo-3-phenylpropionic acid, 3-phenyl-2-oxopropanoic acid, sodium salt
PubChem CID 23667645
IUPAC Name sodium;2-oxo-3-phenylpropanoate
SMILES C1=CC=C(C=C1)CC(=O)C(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 186.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Phenylpyruvate Monohydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.