accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOFFF

CAS RN 114152-23-7

IUPAC Name: 2-(2,3,6-trifluorophenyl)acetic acid
Synonyms: (2,3,6-trifluorophenyl)acetic acid
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: QRAZASHLGLHKEB-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results