Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trifluorophenylacetic acid, 98%

Catalog No. AC426030010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426030010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426030010 Supplier Thermo Scientific Chemicals Supplier No. 426030010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 114152-23-7 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114152-23-7
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00061217
InChI Key QRAZASHLGLHKEB-UHFFFAOYSA-N
Synonym 2,3,6-trifluorophenylacetic acid, 2-2,3,6-trifluorophenyl acetic acid, 2,3,6-trifluorophenyl acetic acid, benzeneacetic acid, 2,3,6-trifluoro, pubchem7293, acmc-1c5f9, rarechem al bo 0509, 2,3,6-trifluorophenylaceticacid, 2,3,6-trifluorophenylaceitc acid, 2,3,6-trifluorobenzeneacetic acid
PubChem CID 2777948
IUPAC Name 2-(2,3,6-trifluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1F)CC(=O)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula F3C6H2CH2COOH
Quantity 1 g
Formula Weight 190.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3, 6-Trifluorophenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.