accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COO

CAS RN 114205-89-9

IUPAC Name: 2-{[2-methyl-4-(9-{3-methyl-4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane
Synonyms: 2-(2-methyl-4-{9-[3-methyl-4-(oxiran-2-ylmethoxy)phenyl]fluoren-9-yl}phenoxymethyl)oxirane
Molecular Weight (g/mol): 490.6
Molecular Formula: C33H30O4
InChi Key: ABXGZTBEQZOCEE-UHFFFAOYNA-N
SMILES: CC1=C(OCC2CO2)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(OCC2CO2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results