Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene 97.0+%, TCI America™
SDP

Catalog No. B46665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46665G 5 g
B46661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46665G Supplier TCI America Supplier No. B46665G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 114205-89-9
Molecular Formula C33H30O4
Molecular Weight (g/mol) 490.60
MDL Number MFCD22494984
InChI Key ABXGZTBEQZOCEE-UHFFFAOYNA-N
Synonym 9,9-Bis(4-hydroxy-3-methylphenyl)fluorene Diglycidyl Ether
PubChem CID 20160952
IUPAC Name 2-{[2-methyl-4-(9-{3-methyl-4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane
SMILES CC1=C(OCC2CO2)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(C)=C(OCC2CO2)C=C1

Specifications

Melting Point 230°C
Color White
Quantity 5 g
Formula Weight 490.60
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-glycidyloxy-3-methylphenyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.