accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 1144-74-7

IUPAC Name: (4-nitrophenyl)(phenyl)methanone
Synonyms: (4-nitrophenyl)(phenyl)methanone
Molecular Weight (g/mol): 227.22
Molecular Formula: C13H9NO3
InChi Key: ZYMCBJWUWHHVRX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)