Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Nitrobenzophenone

Catalog No. N1205825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1205825G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1205825G Supplier Sigma Aldrich Supplier No. N1205825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tris(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite (tris(1,1,1,3,3,3-hexafluoroisopropyl) phosphite) is a sterically hindered, weakly -g-donating and strongly -accepting ligand. it can be prepared by the reaction of lithium salt of 1,1,1,3,3,3-hexafluoro-2-propoxide with pcl3. it reacts with nucleosides to form deoxyribonucleoside 3α-bis(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite units.tris(1,1,1,3,3,3-hexafluoro-2-propyl) phosphite (tris(1,1,1,3,3,3-hexafluoroisopropyl) phosphite) may be used in the following processes:
  • Preparation of new phosphonylating and coupling agents for the synthesis of oligodeoxyribonucleotides
  • Preparation of nucleoside h-phosphonate units
  • Synthesis of tetrakis(1,1,1,3,3,3-hexafluoroisopropoxy)(phenylthio)phosphorane by reacting with 1,1,1,3,3,3-hexafluoroisopropyl benzenesulfenate

Specifications

CAS 1144-74-7
Melting Point 136°C to 138°C (lit.)
Molecular Formula C13H9NO3
Linear Formula O2NC6H4COC6H5
MDL Number MFCD00007354
Quantity 1 g
Molecular Weight (g/mol) 227.22
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.