accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1145-93-3

IUPAC Name: ethoxy ethyl [(4-methoxyphenyl)methyl]phosphonite
Synonyms: ethoxy ethyl (4-methoxyphenyl)methylphosphonite
Molecular Weight (g/mol): 258.25
Molecular Formula: C12H19O4P
InChi Key: XTASVOIVNCSQLW-UHFFFAOYNA-N
SMILES: CCOOP(CC1=CC=C(OC)C=C1)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)