accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114873-10-8

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(2-fluorophenyl)propanoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-(2-fluorophenyl)propanoate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H17FNO4
InChi Key: NTWUXBKUDXGMHV-LLVKDONJSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1F)C([O-])=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)