CAS RN 114873-11-9
CAS RN 114873-11-9
IUPAC Name:
(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-fluorophenyl)propanoic acid
Synonyms:
(2R)-2-[(tert-butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid|BOC-3-FLUORO-D-PHENYLALANINE
Molecular Weight (g/mol):
283.3
Molecular Formula:
C14H18FNO4
InChi Key:
FPCCREICRYPTTL-LLVKDONJSA-N
SMILES:
CC(C)(C)OC(=O)N[C@H](CC1=CC=CC(F)=C1)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Phe 3-F -OH, Advanced ChemTech
CAS: 114873-11-9 Molecular Formula: C14H18FNO4 Molecular Weight (g/mol): 283.30 MDL Number: MFCD00672523 InChI Key: FPCCREICRYPTTL-LLVKDONJSA-N IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-fluorophenyl)propanoic acid SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC(F)=C1)C(O)=O