accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHOHO

CAS RN 115-84-4

IUPAC Name: 2-butyl-2-ethylpropane-1,3-diol
Synonyms: butyl ethyl propanediol
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: DSKYSDCYIODJPC-UHFFFAOYSA-N
SMILES: CCCCC(CC)(CO)CO
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)