Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butyl-2-ethyl-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. B0922500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0922500G 500 g
B092225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0922500G Supplier TCI America Supplier No. B0922500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115-84-4
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.26
MDL Number MFCD00004697
InChI Key DSKYSDCYIODJPC-UHFFFAOYSA-N
Synonym 2-butyl-2-ethyl-1,3-propanediol, 2-ethyl-2-butyl-1,3-propanediol, 3,3-bis hydroxymethyl heptane, 2-butyl-2-ethylpropanediol, caswell no. 129, 1,3-propanediol, 2-butyl-2-ethyl, 2-ethyl-2-butyl-1,3-propandeiol, bepd, unii-61p3kj8k4y, bep
PubChem CID 61038
IUPAC Name 2-butyl-2-ethylpropane-1,3-diol
SMILES CCCCC(CC)(CO)CO

Specifications

Melting Point 43°C
Color White
Boiling Point 178°C
Quantity 500 g
Formula Weight 160.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Butyl-2-ethyl-1,3-propanediol
EINECSNumber (2)-0240&(2)-2391
RTECSNumber TY3605000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.