accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPOOO

CAS RN 115-86-6

IUPAC Name: triphenyl phosphate
Synonyms: triphenyl phosphate phosflex tpp disflamoll TP trifenylfosfat celluflex tpp TPP triphenylphosphate phenyl phosphate ((pho)3PO)
Molecular Weight (g/mol): 326.29
Molecular Formula: C18H15O4P
InChi Key: XZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILES: O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Boiling Point 
  • (2)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)